Order Entry
United Kingdom
ContactUsLinkComponent
Fume hood, benchtop, small, SP Bel-Art
Fume hood, benchtop, small, SP Bel-Art
Catalog # BELAH500000002
undefined
Fume hood, benchtop, small, SP Bel-Art
Catalog # BELAH500000002
Specifications
  • W×D×H ext.:
    690×330×560 mm
  • Description:
    Benchtop, front opening 560×300 mm
  • Pk:
    1
Specifications
About this item
Benchtop fume hood designed for small facilities or installations in which a number of fume hoods are needed, this unit is mainly used in schools, junior colleges and universities.
  • Easy to clean
  • Bottom gap allows for continuous air flow
  • One-piece shell moulded of chemical resistant polyethylene with rounded inside corners
  • Clear acrylic sash slides on a track featuring stop pins for open, closed or intermediate positions