Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Soxhlet extractor
Soxhlet extractor
Supplier:   SMITH SCIENTIFIC
Soxhlet extractor
Supplier:  SMITH SCIENTIFIC
Certificates
Soxhlet extractor
5 Options Available Below
Product Details & Documents
Borosilicate glass.
  • Excellent temperature resistance
  • Excellent chemical resistance
The solvent is recirculated continuously and distilled from the extracted sample before contacting the solid again. The extract is concentrated in the distillation flask. The method allows an effective separation with relatively small amounts of extracting solvents.
Product Family Options
Product Information
  • Socket size
    Availability
    Price
  • SMIT8SEC40/43
    Soxhlet extractor body, 40 ml, borosilicate glass
    Soxhlet extractor body, 40 ml, borosilicate glass
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - £236.00
    £236.00
    1 * 1 items
       £236.00
  • SMIT8SEC60/53
    Soxhlet extractor body, 60 ml, borosilicate glass
    Soxhlet extractor body, 60 ml, borosilicate glass
    34/35 NS
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - £272.00
    £272.00
    1 * 1 items
       £272.00
  • SMIT8SEC60/55
    Soxhlet extractor body, 60 ml, borosilicate glass
    Soxhlet extractor body, 60 ml, borosilicate glass
    34/35 NS
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - £285.00
    £285.00
    1 * 1 items
       £285.00
  • SMIT8SEC100/55
    Soxhlet extractor body, 100 ml, borosilicate glass
    Soxhlet extractor body, 100 ml, borosilicate glass
    34/35 NS
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - £319.00
    £319.00
    1 * 1 items
       £319.00
  • SMIT8SEC200/83
    Soxhlet extractor body, 200 ml, borosilicate glass
    Soxhlet extractor body, 200 ml, borosilicate glass
    50/42 NS
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - £364.00
    £364.00
    1 * 1 items
       £364.00