Dropping funnels, pear shaped, without pressure balance
Supplier: SMITH SCIENTIFIC
Certificates
About this item
Borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Pear shaped, with standard ground joint socket and cone.
Frequently Bought Together