Chemical resistant gloves, Tricopren® 723
Supplier: KCL by Honeywell
Certificates
About this item
This glove is dipped to form the chloroprene layer, then it is dipped in a solvent-free adhesive bath and after that the cotton tricot is pulled on. Thus, the glove is fully coated, impermeable and provides a high chemical and mechanical resistance.
- Free from latex
- Good grip when handling wet parts due to palm profile
- Flexible in warmth an cold, good dexterity
- Seamless tricot allows a very fine touch
Ideal for removal of contaminated parts from dipping baths, handling contaminated heavy parts, cleaning and maintenance in chemical plants and when working with oily and greasy parts.
Certifications: EN 388:2016 (2121X), EN ISO 374-1 Type A (AJKLMS); Cat. III; AQL < 0,65
Food contact approved.
Food contact approved.
Specifications
- Catalog No:
- 112-1751
- 112-1752
- 112-1753
- 112-1754
- Model:
- 723
- 723
- 723
- 723
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Liner material:
- Cotton
- Cotton
- Cotton
- Cotton
- Cuff style:
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet
- EN norm:
- EN 388:2016 (2121X), EN ISO 374-1 Type A (AJKLMS)
- EN 388:2016 (2121X), EN ISO 374-1 Type A (AJKLMS)
- EN 388:2016 (2121X), EN ISO 374-1 Type A (AJKLMS)
- EN 388:2016 (2121X), EN ISO 374-1 Type A (AJKLMS)
- Food contact approved:
- +
- +
- +
- +