Counting chamber, Helber
720-0105
- Depth of chamber:0,02 mm
- Type:Thomas
- Description:Counting chamber, Helber
- Pk:1
The Helber counting chamber ensures durability and accuracy.
- Thoma ruling on a single round plateau
Specially designed for cell counting applications.