Order Entry
United Kingdom
ContactUsLinkComponent
Anion exchange HPLC columns, Dionex™, IonPac™ AS11
Anion exchange HPLC columns, Dionex™, IonPac™ AS11
Supplier:  DIONEX
Certificates
Anion exchange HPLC columns, Dionex™, IonPac™ AS11
Supplier:  DIONEX
Specifications
  • Catalog No:
  • DION044076
  • DION044078
  • Max. pressure:
  • 4000 psi
  • 4000 psi
  • pH range:
  • 0 - 14
  • 0 - 14