Pliers for stainless steel tubing
554-1105
- Description:Tubing pliers, for cutting 1/16" stainless steel tubing off a coil
- Pk:1
The pliers type of cutter can be used for quick cuts for non-critical applications or at hard to get places.
- For easy cutting of ¹/₁₆" tubing off a coil