Surface cleansers, Greenline Plus
Catalog # BUNZ033425
Supplier: BUNZL
Specifications
- Description:Greenline Plus Hard surface cleaner RTU
- Packed:750 ml
- Pk:6
Specifications
About this item
Liquid cleanser in spray bottle for cleaning floors and hard surfaces in offices, schools, hospitals, canteens etc. Suitable for use on linoleum, PVC, rubber, sealed wood, sealed cork, thermoplastic floors, most modern hard surfaces.
Caution: Not recommended for laminated flooring or unsealed wood and cork.