Order Entry
United Kingdom
ContactUsLinkComponent
Absorption granules for body fluid spills, Haz-Tab
Absorption granules for body fluid spills, Haz-Tab
Catalog # GUESH8800
Supplier:  GUEST
Absorption granules for body fluid spills, Haz-Tab
Catalog # GUESH8800
Supplier:  GUEST
Specifications
  • Description:
    Haz-Tab granules, with NaDCC, for the absorption of blood and blood stained body fluid spills
  • Seq:
    01
  • Packed:
    500 g/shaker
  • Pk:
    500 g
Specifications
About this item
A good alternative method to using hypochlorite to safely deal with blood or blood contaminated spillages.
The granules contain and absorb the spill and also incorporate a strong disinfectant NaDCC which is chlorine releasing.