Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Combination pH electrodes, general purpose
Marketsource
Supplier:   Beckman Coulter
Combination pH electrodes, general purpose
Supplier:  Beckman Coulter
Marketsource
Certificates
Combination pH electrodes, general purpose
Combination pH electrodes, general purpose
3 Options Available Below
Product Details & Documents
Combination pH electrodes for general purpose pH measurements.
  • 1 m long integrated cable with BNC connector
  • Glass or epoxy body
  • Epoxy body has a protective design in which body extend over glass bulb measuring surface
Product Family Options
Product Information
  • Type
    pH
    Temp.
    Material
    Electrolyte
    Ø×L
    Connection
    For
    Availability
    Price
  • Semi-micro
    0 - 14
    0...+100 °C
    Epoxy
    3,5M KCl (refillable)
    6×150 mm
    BNC, 1 m cable
    General purposes
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    BECLA57187
    Marketsource
  • Standard
    0 - 14
    0...+100 °C
    Epoxy
    3,5M KCl (refillable)
    12×150 mm
    BNC, 1 m cable
    General purposes
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    BECLA57186
    Marketsource
  • Semi-micro
    0 - 14
    0...+100 °C
    Glass
    3,5M KCl (refillable)
    6×150 mm
    BNC, 1 m cable
    General purposes
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    BECLA57183
    Marketsource