
2-Amino-6-chloropyrazine 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:33332-28-4
:
Formula:C₄H₄ClN₃
MDL Number:MFCD00055024
:129.55 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together