Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
2-Chlorophenol

2-Chlorophenol

Glass bottle for solids, 1 kg
Catalog # MOLE23133552-1KG
Irritant
Environment
Marketsource
Supplier:   Molekula
2-Chlorophenol
Catalog # MOLE23133552-1KG
Supplier:  Molekula
Marketsource
CAS Number:  95-57-8
2-Chlorophenol

Glass bottle for solids, 1 kg

CAS Number:  95-57-8
Price
Inquire For Price
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  1-Chloro-2-hydroxybenzene
  • CAS Number:95-57-8
  • Formula:C₆H₅ClO
  • MDL Number:MFCD00002159
  • Molecular Weight:128.56 g/mol
  • Merck Index:12,02173
  • Boiling Pt:175 °C (1013 hPa)
  • Melting Pt:9 °C
  • Density:1.25 g/cm³ (20 °C)
  • Flash Pt:72 °C
  • Storage Temperature:Ambient
  • EINECS:202-433-2
  • UN:2021
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications