3-Chloro-2-fluoro-N-methylaniline hydrochloride
Catalog # COBBHC-2858-1G
Supplier: COMBI-BLOCKS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
CAS Number:1187386-17-9
Formula:C₇H₈Cl₂FN
MDL Number:MFCD12913956