3-(2-Chlorophenoxy)propionic acid
Glass bottle for solids, 1 g
Catalog # ACTIAS22928G1
Marketsource
Supplier: ACTIVATE SCIENTIFIC
3-(2-Chlorophenoxy)propionic acid
Catalog # ACTIAS22928G1
Supplier: ACTIVATE SCIENTIFIC
Marketsource
CAS Number: 7170-45-8
Jump to:
Product Details & Documents
Synonyms: 3-(2-Chlorophenoxy)propanoic acid
- CAS Number:7170-45-8
- Formula:C₉H₉ClO₃
- MDL Number:MFCD02295728
- Molecular Weight:200.6190 g/mol
- EINECS:230-521-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



