N(α)-Cbz-DL-tryptophan
Catalog # BACE4011146.0025
Supplier: BACHEM BIOCHEMICA
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
Specifications
About this item
Synonyms: Cbz-DL-Trp-OH, Z-DL-Trp-OH
:
:338.4 g/mol
EINECS:235-949-1
CAS Number:13058-16-7
Formula:C₁₉H₁₈N₂O₄
MDL Number:MFCD00069707