Chromium(III) picolinate, Sigma-Aldrich®
Catalog # SIALC4124-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
CAS Number:14639-25-9
Formula:C₁₈H₁₂CrN₃O₆
MDL Number:MFCD00068715