Order Entry
United Kingdom
ContactUsLinkComponent
Nordihydroguaiaretic acid, Millipore®
Nordihydroguaiaretic acid, Millipore®
Catalog # 479975-250
CAS Number:  
Nordihydroguaiaretic acid, Millipore®
Catalog # 479975-250
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :302.37 g/mol
EINECS:207-903-0
CAS Number:500-38-9
Formula:C₁₈H₂₂O₄
MDL Number:MFCD00002206