Ethyl-2-amino-2-cyanoacetate mono(4-methylbenzenesulphonate)
Catalog # COBBSS-3239-1G
Supplier: COMBI-BLOCKS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:300.3308 g/mol
CAS Number:37842-58-3
Formula:C₁₂H₁₆N₂O₅S
MDL Number:MFCD00666813