N-Formyl 2-chloro-5-(trifluoromethyl)aniline ≥96%
Supplier: COMBI-BLOCKS
Certificates
About this item
Synonyms: N-Formyl 2-chloro-5-(trifluoromethyl)aniline, 6'-CHLORO-ALPHA,ALPHA,ALPHA-TRIFLUORO-M-FORMOTOLUIDIDE, N-(2-Chloro-5-(trifluoromethyl)phenyl)formamide
CAS Number:657-63-6
:
Formula:C₈H₅ClF₃NO
MDL Number:MFCD00021000
Molecular Weight:223.6 g/mol
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together