
3-(Methylsulphonyl)aniline hydrochloride 95%
ACRO424870025
:
- Pack type:Glass bottle
- Pk:2,5 g
- Seq:3122
- Appearance:Off-white to light beige Powder
- Infrared spectrum:Conforms
- Titration Argentometric:≥94 %
- Loss on drying:≤5 %
:
:227…230 °C
:GHS07
:Ambient
:80213-28-1
:C₇H₁₀ClNO₂S
:MFCD00216487