3-Chloro-5-(tert-butyldimethylsilyloxy)phenylboronic acid pinacol ester
Supplier: COMBI-BLOCKS
Certificates
About this item
CAS Number:1218789-46-8
:
Formula:C₁₈H₃₀BClO₃Si
MDL Number:MFCD12546583
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together