Lithium phenoxide 1 mol/l in THF, Sigma-Aldrich®
Catalog # SIAL400378-100ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
EINECS:209-086-6
CAS Number:555-24-8
Formula:C₆H₅LiO
MDL Number:MFCD00013473