1-Pyrenecarboxylic acid, Sigma-Aldrich®
Catalog # SIAL391581-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:246.2601 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:19694-02-1
Formula:C₁₇H₁₀O₂
MDL Number:MFCD00192424