Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Methyl-1H-pyrazol-4-carbaldehyde
1-Methyl-1H-pyrazol-4-carbaldehyde 96%
1-Methyl-1H-pyrazol-4-carbaldehyde 96%
1-Methyl-1H-pyrazol-4-carbaldehyde 96%
H54146.03
:
Thermo Fisher Scientific
:
1-Methyl-1H-pyrazol-4-carbaldehyde 96%
H54146.03
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle for solids
Pk:
1 g
:
25016-11-9
:
:
C₅H₆N₂O
:
MFCD00460465
:
110.12 g/mol
:
29…34 °C
:
>230 °C (446 °F)
:
GHS05
:
GHS07
:
Ambient