2-(Tributylstannyl)pyrazine 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:205371-27-3
:
Formula:C₁₆H₃₀N₂Sn
MDL Number:MFCD01319077
Boiling Pt:130 °C (1 mmHg)
Density:1.171
Flash Pt:>110 °C (230 °F)
ADR:6.1,III
:369.12 g/mol
:GHS06
:GHS08
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together