6-(Trifluoromethyl)nicotinamide
Catalog # ENAMEN300100828.1G
Supplier: SIA ENAMINE
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:190.1200 g/mol
CAS Number:386715-35-1
Formula:C₇H₅F₃N₂O
MDL Number:MFCD01862646