Order Entry
United Kingdom
ContactUsLinkComponent
Methyl orange 0.1% in ethanol 20% (v/v) Reag. Ph. Eur.
Methyl orange 0.1% in ethanol 20% (v/v) Reag. Ph. Eur.
Catalog # REGE1054802
Supplier:  REAGECON
CAS Number:  
Methyl orange 0.1% in ethanol 20% (v/v) Reag. Ph. Eur.
Catalog # REGE1054802
Supplier:  REAGECON
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :327.34 g/mol
EINECS:208-925-3
UN:1170
ADR:3,III
CAS Number:547-58-0
Formula:C₁₄H₁₄N₃NaO₃S
MDL Number:MFCD00007502