
3-Amino-4-bromo-1H-indazole 97%
Catalog # H33460.MD
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
Specifications
About this item
:
:212.05 g/mol
Melting Pt:142…144 °C
:GHS07
Storage Temperature:Ambient
CAS Number:914311-50-5
Formula:C₇H₆BrN₃
MDL Number:MFCD09909580