3-(3-Bromophenyl)-1H-pyrazole
Supplier: COMBI-BLOCKS
Certificates
About this item
Synonyms: 3-(3-Bromophenyl)-1H-pyrazole
CAS Number:948294-12-0
:149739-65-1
Formula:C₉H₇BrN₂
MDL Number:MFCD01940433
:223.07 g/mol
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together