2-(2-Thienyl)isonicotinic acid
Catalog # COBBYA-5955-1G
Supplier: COMBI-BLOCKS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
CAS Number:893723-57-4
Formula:C₁₀H₇NO₂S
MDL Number:MFCD06496472