Order Entry
United Kingdom
ContactUsLinkComponent
3-Bromo-1H-pyrazolo[4,3-b]pyridine
3-Bromo-1H-pyrazolo[4,3-b]pyridine
Catalog # ACTIAS2515G1
CAS Number:  
3-Bromo-1H-pyrazolo[4,3-b]pyridine
Catalog # ACTIAS2515G1
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :198.0201 g/mol
CAS Number:633328-33-3
Formula:C₆H₄BrN₃
MDL Number:MFCD11518974