


2-(4-Morpholino)-5-(tri-n-butylstannyl)pyridine
Catalog # ACTIAS51220G1
Supplier: ACTIVATE SCIENTIFIC
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:GHS06
:GHS08
:GHS09
CAS Number:223556-07-8
Formula:C₂₁H₃₈N₂OSn
MDL Number:MFCD09025804