Methyl-6-bromo-7-methylimidazo[1,2-a]pyridine-2-carboxylate
Catalog # COBBPY-7112-1G
Supplier: COMBI-BLOCKS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
CAS Number:907945-87-3
Formula:C₁₁H₁₁BrN₂O₂
MDL Number:MFCD09475851