Industrial gloves, Monsun® 105
These tight-fitting knitted cotton liners with palm nitrile coating and knit wrist are ideal for precision jobs.
- Moisture-repellent, very breathable
- Glove surface free from silicone (lacquer indifference, test method automotive industry)
- Particularly close-fitting for applications where sensitivity is needed
- Allows longer working without showing signs of fatigue
- Additional wrist protection
Suitable applications include assembly and transportation works, manufacturing of small parts, warehouse and packaging applications, metal processing, precision engineering and electronical as well as automotive industries.
Thickness: 0,75 ± 0,05 mm
Colour: Yellow/white
EN 388 (2111), Cat. II Food contact approved
: EN 388:2016 (2111X), Cat. II
Food contact approved
Food contact approved
- Catalog No:
- 112-1227
- 112-1228
- 112-1225
- 112-1226
- Model:
- 105
- 105
- 105
- 105
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Cotton
- Cotton
- Cotton
- Cotton
- Finish:
- ¾ coating
- ¾ coating
- ¾ coating
- ¾ coating
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (2111X)
- EN 388:2016 (2111X)
- EN 388:2016 (2111X)
- EN 388:2016 (2111X)
- Food contact approved:
- +
- +
- +
- +
Frequently Bought Together