Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Anisaldehyde solutions
Anisaldehyde solution Reag. Ph. Eur. 1007301
Anisaldehyde solution Reag. Ph. Eur. 1007301
87777.180
:
VWR Chemicals
:
Anisaldehyde solution Reag. Ph. Eur. 1007301
87777.180
:
VWR Chemicals
:
Pack type:
Plastic bottle
Pk:
100 mL
Seq:
2440
Environmentally Preferable:
:
:
2924
:
GHS02
:
GHS06
:
GHS05
:
GHS08
:
3 (8) II