Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Methyl-3,4-dimethylbenzoate ≥98%

Methyl-3,4-dimethylbenzoate ≥98%

Glass bottle for solids, 250 g
Catalog # ABCRAB125607.0250
Marketsource
Supplier:   abcr
Methyl-3,4-dimethylbenzoate ≥98%
Catalog # ABCRAB125607.0250
Supplier:  abcr
Marketsource
CAS Number:  38404-42-1
Methyl-3,4-dimethylbenzoate ≥98%

Glass bottle for solids, 250 g

CAS Number:  38404-42-1
Price
Inquire For Price
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 98% CAS No.: 38404-42-1 Molecular Formula: C10H12O2 Formula Weight: 164.21 Melting Point: 35-37° MDL No.: MFCD00219861

Synonyms:  Methyl 3,4-dimethylbenzoate
  • CAS Number:38404-42-1
  • Formula:C₁₀H₁₂O₂
  • MDL Number:MFCD00219861
  • Molecular Weight:164.2011 g/mol
  • Melting Pt:35…37 °C
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications