Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(4-Fluorophenyl)-3-thiosemicarbazide
4-(4-Fluorophenyl)-3-thiosemicarbazide ≥98%
4-(4-Fluorophenyl)-3-thiosemicarbazide ≥98%
4-(4-Fluorophenyl)-3-thiosemicarbazide ≥98%
L09482.06
:
Thermo Fisher Scientific
:
4-(4-Fluorophenyl)-3-thiosemicarbazide ≥98%
L09482.06
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
Environmentally Preferable:
:
:
185.22 g/mol
:
∼ 179 °C (dec.)
:
6.1,III
:
GHS06
:
Ambient
:
330-94-9
:
C₇H₈FN₃S
:
MFCD00041294