


Ethylamine 2.0 M in THF
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75-04-7
:
Formula:C₂H₇N
MDL Number:MFCD00008160
:45.08 g/mol
Density:0.856
Flash Pt:–34 °C (–29 °F)
EINECS:200-834-7
UN:2056
ADR:3,II
:GHS02
:GHS06
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together