3-Cyano-4-fluorobenzenesulphonyl chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:351003-23-1
:
Formula:C₇H₃ClFNO₂S
MDL Number:MFCD03093760
:219.62 g/mol
ADR:8,I
:M-Clarity™ quality level = MQ200
UN:3261
Frequently Bought Together