Order Entry
United Kingdom
ContactUsLinkComponent
2-(Ethoxycarbonyl)furan-5-boronic acid pinacol ester
2-(Ethoxycarbonyl)furan-5-boronic acid pinacol ester
Catalog # COBBPN-5514-1G
Supplier:  COMBI-BLOCKS
CAS Number:  
2-(Ethoxycarbonyl)furan-5-boronic acid pinacol ester
Catalog # COBBPN-5514-1G
Supplier:  COMBI-BLOCKS
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Ethyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)furan-2-carboxylate
 :
 :266.10 g/mol
CAS Number:1073338-92-7
Formula:C₁₃H₁₉BO₅
MDL Number:MFCD11855980