4,4'-Dihydroxybenzophenone
Glass bottle for solids, 100 g
Catalog # ACTIAS21999G100
Marketsource
Supplier: ACTIVATE SCIENTIFIC
4,4'-Dihydroxybenzophenone
Catalog # ACTIAS21999G100
Supplier: ACTIVATE SCIENTIFIC
Marketsource
CAS Number: 611-99-4
Jump to:
Product Details & Documents
Synonyms: Bis(4-hydroxyphenyl) ketone
- CAS Number:611-99-4
- Formula:C₁₃H₁₀O₃
- MDL Number:MFCD00002358
- Molecular Weight:214.22 g/mol
- Boiling Pt:375 °C (1013 hPa)
- Melting Pt:214…217 °C
- Density:1,336 g/cm³ (20 °C)
- EINECS:210-288-1
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



