2'-Hydroxyacetophenone organic reference standard
Catalog # PROOMM0042.13
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:136.15 g/mol
Boiling Pt:213 °C (1013 hPa)
Density:1.13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-288-0
CAS Number:118-93-4
:MFCD00002219
Formula:C₈H₈O₂