Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-(4-Methyl-2-pyridyl)thiourea
N-(4-Methyl-2-pyridyl)thiourea
N-(4-Methyl-2-pyridyl)thiourea
N-(4-Methyl-2-pyridyl)thiourea
FLUO020744-1G
:
FLUOROCHEM
:
N-(4-Methyl-2-pyridyl)thiourea
FLUO020744-1G
:
FLUOROCHEM
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
167.23 g/mol
:
21242-21-7
:
C₇H₉N₃S
:
MFCD04117348