Trimethylamine 25% (w/w) in propylene glycol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:75-50-3
:
Formula:C₃H₉N
MDL Number:MFCD00008327
:M-Clarity™ quality level = MQ300
:59.11 g/mol
EINECS:200-875-0
Frequently Bought Together