Order Entry
United Kingdom
ContactUsLinkComponent
5-Fluoro-2-methylbenzaldehyde, Sigma-Aldrich®
5-Fluoro-2-methylbenzaldehyde, Sigma-Aldrich®
Catalog # SIAL685003-1G
Supplier:  Merck
CAS Number:  
5-Fluoro-2-methylbenzaldehyde, Sigma-Aldrich®
Catalog # SIAL685003-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ100
 :138.14 g/mol
CAS Number:22062-53-9
Formula:C₈H₇FO
MDL Number:MFCD02261763