5-Fluoro-2-methylbenzaldehyde
Catalog # FLUO022378-1G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:138.14 g/mol
CAS Number:22062-53-9
Formula:C₈H₇FO
MDL Number:MFCD02261763