5-Fluoro-2-methylbenzaldehyde
Catalog # ABCRAB144145.0005
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
About this item
:
:138.14 g/mol
CAS Number:22062-53-9
Formula:C₈H₇FO
MDL Number:MFCD02261763