4,5-Dimethoxy-2-nitrotoluene
Catalog # FLUO018589-5G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:197.187 g/mol
CAS Number:7509-11-7
Formula:C₉H₁₁NO₄
MDL Number:MFCD00060708