(±)-Naringenin, Sigma-Aldrich®
: Merck
:67604-48-2
:93602-28-9
:C₁₅H₁₂O₅
:MFCD00006844
:272.25 g/mol
:266-769-1
Frequently Bought Together
Ethyl acetate ≥99.5% for HPLC
: Thermo Fisher Scientific
1-Propanol ≥99%, GPR RECTAPUR®
: VWR Chemicals
Hydrochloric acid ≥36.5% ≥99.999% (metals basis)
: Thermo Fisher Scientific
Methanol, extra dry 99.9%, AcroSeal®
: Thermo Fisher Scientific
Acetone ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Dimethyl sulphoxide ≥99.9% ACS, Riedel-de Haën™
: Honeywell Chemicals
2-Butoxyethanol 99%, extra pure
: Thermo Fisher Scientific
N,N-Dimethylacetamide ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
: Honeywell Chemicals
Chloroform ≥99.5% stabilised
: Honeywell Chemicals
Toluene 99.8+% for analysis
: Thermo Fisher Scientific
Isobutanol 99+% for HPLC
: Thermo Fisher Scientific
Diisopropyl ether stabilised for analysis
: Thermo Fisher Scientific
Ethanol absolute Ph. Eur., J.T.Baker®
: Avantor