2-Piperazinonicotinonitrile
Catalog # MAYBHTS04470DA
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
CAS Number:84951-44-0
Formula:C₁₀H₁₂N₄
MDL Number:MFCD00190248